Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse LIF Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Supplier: R&D Systems AB449NA
This item is not returnable.
View return policy
Description
LIF Polyclonal specifically detects LIF in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.Specifications
LIF | |
Polyclonal | |
Unconjugated | |
CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDA, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI | |
Goat | |
Protein A or G purified | |
RUO | |
3976 | |
Reconstitute at 1 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG |
Western Blot, Immunohistochemistry, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 80-480 ng/mL | |
LIF | |
E. coli-derived recombinant mouse LIF | |
1 mg | |
Primary | |
Detects mouse LIF in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human LIF is observed. Neutralizes the biological activity of recombinant mouse LIF and will also neutralize the biological activity of recombinant human LIF at a 10-fold higher IgG concentration. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction