Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse LIGHT/TNFSF14 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB17943S100UG
This item is not returnable.
View return policy
Description
LIGHT/TNFSF14 Monoclonal specifically detects LIGHT/TNFSF14 in Mouse samples. It is validated for Flow Cytometry.Specifications
LIGHT/TNFSF14 | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/ml in a saline solution containing BSA and Sodium Azide. | |
CD258, CD258 antigen, delta transmembrane LIGHT, Herpes virus entry mediator ligand, Herpesvirus entry mediator ligand, HVEM-L, HVEMLherpesvirus entry mediator-ligand, ligand for herpesvirus entry mediator, LIGHTherpesvirus entry mediator A, LTg, TNFSF14, TR2, tumor necrosis factor (ligand) superfamily, member 14, tumor necrosis factor ligand superfamily member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT | |
Mouse myeloma cell line NS0-derived recombinant mouse LIGHT/TNFSF14 Asp72-Val239 Accession # Q9QYH9 | |
100 μg | |
8740 | |
Mouse | |
Supernatant |
Flow Cytometry | |
906909 | |
Flow Cytometry 0.25-1 μg/10^6 cells | |
Q9QYH9 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects mouse LIGHT/TNFSF14 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction