Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse LIGHT/TNFSF14 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
$200.50 - $573.00
Specifications
Antigen | LIGHT/TNFSF14 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
LIGHT/TNFSF14 Polyclonal specifically detects LIGHT/TNFSF14 in Mouse samples. It is validated for Western Blot.Specifications
LIGHT/TNFSF14 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
CD258, CD258 antigen, delta transmembrane LIGHT, Herpes virus entry mediator ligand, Herpesvirus entry mediator ligand, HVEM-L, HVEMLherpesvirus entry mediator-ligand, ligand for herpesvirus entry mediator, LIGHTherpesvirus entry mediator A, LTg, TNFSF14, TR2, tumor necrosis factor (ligand) superfamily, member 14, tumor necrosis factor ligand superfamily member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT | |
TNFSF14 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9QYH9 | |
8740 | |
Mouse myeloma cell line NS0-derived recombinant mouse LIGHT (R&D Systems, Catalog # 1794-LT) Asp72-Val239 Accession # Q9QYH9 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse LIGHT in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human LIGHT is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title