Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse LIGHT/TNFSF14 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF1794
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF1794 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF1794 Supplier R&D Systems Supplier No. BAF1794
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

LIGHT/TNFSF14 Polyclonal specifically detects LIGHT/TNFSF14 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen LIGHT/TNFSF14
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q9QYH9
Gene Alias CD258, CD258 antigen, delta transmembrane LIGHT, Herpes virus entry mediator ligand, Herpesvirus entry mediator ligand, HVEM-L, HVEMLherpesvirus entry mediator-ligand, ligand for herpesvirus entry mediator, LIGHTherpesvirus entry mediator A, LTg, TNFSF14, TR2, tumor necrosis factor (ligand) superfamily, member 14, tumor necrosis factor ligand superfamily member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT
Gene Symbols TNFSF14
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse LIGHT/TNFSF14 Asp72-Val239 Accession # Q9QYH9
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8740
Test Specificity Detects mouse LIGHT/TNFSF14 in Western blots. In Western blots, approximately 50% cross-reactivity with recombinant human LIGHT is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.