Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse LILRC1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4334
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4334 100 μg
AF4334SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF4334 Supplier R&D Systems Supplier No. AF4334
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

LILRC1 Polyclonal specifically detects LILRC1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen LILRC1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. AAY44811
Gene Symbols Lilra5
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse LILRC1 Gln17-Asn248 Accession # AAY44811
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 232801
Test Specificity Detects mouse LILRC1 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant rat (rr) LILRC1 is observed and approximately 15% cross-reactivity with rrLILRC2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.