Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse LILRC1 Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems BAF4334
This item is not returnable.
View return policy
Description
LILRC1 Polyclonal specifically detects LILRC1 in Mouse samples. It is validated for Western Blot.Specifications
| LILRC1 | |
| Polyclonal | |
| Biotin | |
| AAY44811 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 232801 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Lilra5 | |
| Mouse myeloma cell line NS0-derived recombinant mouse LILRC1 Gln17-Asn248 Accession # AAY44811 | |
| 50 μg | |
| Primary | |
| Detects mouse LILRC1 in Western blots. In Western blots, approximately 30% cross-reactivity with recombinant rat (rr) LILRC1 is observed and approximately 10% cross-reactivity with rrLILRC2 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction