Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Limitin/IFN-zeta Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB597SP
This item is not returnable.
View return policy
Description
Limitin/IFN-zeta Monoclonal specifically detects Limitin/IFN-zeta in Mouse samples. It is validated for Western Blot.Specifications
Limitin/IFN-zeta | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
6030405N23Rik, BGIF, Ifnz, IFN-z, IFNzeta, interferon zeta, Lmtn | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
319146 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot | |
183727 | |
Western Blot 1 ug/mL | |
Q9R1T0 | |
Ifnz | |
Mouse myeloma cell line NS0-derived recombinant mouse Limitin/IFN-zeta Leu22-Arg182 Accession # Q9R1T0 | |
25 μg | |
Primary | |
Detects mouse Limitin/IFN-zeta in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse IFN-gamma is observed. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction