Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Limitin/IFN-zeta Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $643.50
Specifications
Antigen | Limitin/IFN-zeta |
---|---|
Clone | 183707 |
Dilution | Neutralization 0.02-0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Neutralization, ELISA |
Classification | Monoclonal |
Description
Limitin/IFN-zeta Monoclonal specifically detects Limitin/IFN-zeta in Mouse samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
Limitin/IFN-zeta | |
Neutralization 0.02-0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
6030405N23Rik, BGIF, Ifnz, IFN-z, IFNzeta, interferon zeta, Lmtn | |
Ifnz | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
183707 | |
Neutralization, ELISA | |
Unconjugated | |
Rat | |
Mouse | |
Q9R1T0 | |
319146 | |
Mouse myeloma cell line NS0-derived recombinant mouse Limitin/IFN-zeta Leu22-Arg182 Accession # Q9R1T0 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Limitin/IFN-zeta in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse IFN-gamma is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title