Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Limitin/IFN-zeta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF597
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF597 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF597 Supplier R&D Systems Supplier No. AF597
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Limitin/IFN-zeta Polyclonal specifically detects Limitin/IFN-zeta in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.

Specifications

Antigen Limitin/IFN-zeta
Applications Western Blot, Immunohistochemistry, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 1.5-7.5 ng/mL
Gene Accession No. Q9R1T0
Gene Alias 6030405N23Rik, BGIF, Ifnz, IFN-z, IFNzeta, interferon zeta, Lmtn
Gene Symbols Ifnz
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Limitin/IFN-zeta Leu22-Arg182 Accession # Q9R1T0
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 319146
Test Specificity Detects Limitin/IFN-zeta in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse IFN-gamma is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.