Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Lipocalin-2/NGAL Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF1857SP
This item is not returnable.
View return policy
Description
Lipocalin-2/NGAL Polyclonal specifically detects Lipocalin-2/NGAL in Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation.Specifications
Lipocalin-2/NGAL | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative | |
24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin2, Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGAL, NGALlipocalin-2, Oncogene 24p3, p25, Siderocalin, Uterocalin | |
Goat | |
Affinity Purified | |
RUO | |
3934 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
LYOPH | |
Western Blot 0.25 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 3-15 ug/mL, Immunoprecipitation 25 ug/mL | |
P11672 | |
LCN2 | |
Mouse myeloma cell line NS0-derived recombinant mouse Lipocalin-2/NGAL Gln21-Asn200 Accession # P11672 | |
25 μg | |
Primary | |
Detects mouse Lipocalin-2/NGAL in direct ELISAs and Western blots. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction