Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Lipocalin-2/NGAL Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB18571500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB18571500 500 μg
MAB18571SP 25 μg
MAB18571100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB18571500 Supplier R&D Systems Supplier No. MAB18571500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Lipocalin-2/NGAL Monoclonal specifically detects Lipocalin-2/NGAL in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Lipocalin-2/NGAL
Applications ELISA
Classification Monoclonal
Clone 228421
Concentration LYOPH
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P11672
Gene Alias 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin2, Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGAL, NGALlipocalin-2, Oncogene 24p3, p25, Siderocalin, Uterocalin
Gene Symbols LCN2
Host Species Rat
Immunogen NS0-derived recombinant mouse Lipocalin-2/NGAL Gln21-Asn200 Accession # P11672
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3934
Test Specificity Detects mouse Lipocalin-2/NGAL in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant human (rh) Lipocalin-1, rhLipocalin-2 or recombinant rat Lipocalin-2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.