Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse LIX Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB433100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB433100 100 μg
MAB433SP 25 μg
MAB433500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB433100 Supplier R&D Systems Supplier No. MAB433100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 9 publications

LIX Monoclonal specifically detects LIX in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen LIX
Applications Western Blot, Neutralization, ELISA
Classification Monoclonal
Clone 61905
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.5-2.5 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P50228
Gene Symbols Cxcl5
Host Species Rat
Immunogen E. coli-derived recombinant mouse LIX Val45-Ala118 Accession # P50228
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6374
Test Specificity Detects mouse LIX in Western blots. In Western blots, no cross-reactivity with recombinant human CXCL1, 2 3 5 6 7 8 9, 10, 11, 12/SDF-1 alpha , 12/SDF-1 beta , 13, recombinant mouse CXCL1, 2, 6, 9, 10, 12/SDF-1 alpha , 13, recombinant rat CXCL1, 3/CINC-2 alpha , 3/CINC-2 beta , or recombinant porcine CXCL8 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.