Learn More
Description
Specifications
Specifications
| Product Type | ELISA Development Kit |
| Quantity | 15 x 96-well Plates |
| Species | Mouse |
| Conjugate | Biotin |
| Assay Range | 15.60 to 1000 pg/mL |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 20311 |
| Product Line | R&D Systems |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Research Area | Chemokines |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
