Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse LOX-1/OLR1 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB1564R100UG
This item is not returnable.
View return policy
Description
LOX-1/OLR1 Monoclonal specifically detects LOX-1/OLR1 in Mouse samples. It is validated for Flow Cytometry.Specifications
LOX-1/OLR1 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
AAG44998 | |
OLR1 | |
Mouse myeloma cell line NS0-derived recombinant mouse LOX-1 Arg60-Ile363 Accession # AAG44998 | |
RUO | |
4973 | |
Mouse | |
IgG2a |
Flow Cytometry | |
214012 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CLEC8A, CLEC8ASLOX1, C-type lectin domain family 8 member A, hLOX-1, Lectin-like oxidized LDL receptor 1, Lectin-like oxLDL receptor 1, Lectin-type oxidized LDL receptor 1, LOX1, LOX-1, LOX1ox LDL receptor 1, LOXIN, OLR1, oxidised low density lipoprotein (lectin-like) receptor 1, oxidized low density lipoprotein (lectin-like) receptor 1, oxidized low-density lipoprotein receptor 1, oxidized low-density lipoprotein receptor 1, soluble form, Ox-LDL receptor 1, SCARE1, scavenger receptor class E, member 1, SR-E1 | |
Rat | |
100 μg | |
Primary | |
Detects mouse LOX-1 in direct ELISAs and Western blots. In these formats, this antibody does not cross-react with recombinant human LOX-1. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction