Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse LOX-1/OLR1 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $2020.00
Specifications
Antigen | LOX-1/OLR1 |
---|---|
Clone | 214012 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1564SP
![]() |
R&D Systems
MAB1564SP |
25 μg |
Each for $200.50
|
|
|||||
MAB1564100
![]() |
R&D Systems
MAB1564100 |
100 μg |
Each for $573.00
|
|
|||||
MAB1564500
![]() |
R&D Systems
MAB1564500 |
500 μg |
Each for $2,020.00
|
|
|||||
Description
LOX-1/OLR1 Monoclonal specifically detects LOX-1/OLR1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
LOX-1/OLR1 | |
LYOPH | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CLEC8A, CLEC8ASLOX1, C-type lectin domain family 8 member A, hLOX-1, Lectin-like oxidized LDL receptor 1, Lectin-like oxLDL receptor 1, Lectin-type oxidized LDL receptor 1, LOX1, LOX-1, LOX1ox LDL receptor 1, LOXIN, OLR1, oxidised low density lipoprotein (lectin-like) receptor 1, oxidized low density lipoprotein (lectin-like) receptor 1, oxidized low-density lipoprotein receptor 1, oxidized low-density lipoprotein receptor 1, soluble form, Ox-LDL receptor 1, SCARE1, scavenger receptor class E, member 1, SR-E1 | |
OLR1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
214012 | |
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Rat | |
Mouse | |
AAG44998 | |
4973 | |
Mouse myeloma cell line NS0-derived recombinant mouse LOX-1 Arg60-Ile363 Accession # AAG44998 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse LOX-1 in direct ELISAs and Western blots. In these formats, this antibody does not cross-react with recombinant human LOX-1. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title