Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse LOX-1/OLR1 ELISA Kit

Catalog No. EM50RBX10
Encompass_Preferred
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests

ELISA

Mouse LOX-1/OLR1 quantitates mouse LOX-1/OLR1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse LOX-1/OLR1.

LOX-1 (lectin-like oxidized LDL receptor -1) is a 40-50 kDa type II membrane glycoprotein class E scavenger receptor expressed by endothelial cells, macrophages, activated platelets and smooth muscle cells. LOX-1 binds to a wide range of ligands, including oxidized low-density lipoprotein (oxLDL), hypochlorite modified high-density lipoprotein (HDL), aged /apoptotic cells, activated platelets and bacteria, reflecting its versatile physiological functions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number CLEC8A; C-type lectin domain family 8 member A; hLOX 1; hLO x 1; hLOX-1; lectin-like oxidized LDL receptor; lectin-like oxidized LDL receptor 1; lectin-like oxidized low-density lipoprotein receptor; Lectin-like oxidized low-density lipoprotein receptor-1; lectin-like oxLDL receptor 1; lectin-type oxidized LDL receptor 1; LOX 1; LO x 1; LOX-1; LOXIN; Oldlr1; Oldr1; Olr1; ox LDL receptor 1; oxidised low density lipoprotein (lectin-like) receptor 1; oxidized low density lipoprotein (lectin-like) receptor 1; oxidized low density lipoprotein receptor 1; oxidized low-density lipoprotein receptor 1; Oxidized low-density lipoprotein receptor 1, soluble form; ox-LDL receptor 1; SCARE1; scavenger receptor class E, member 1; sLOX 1; SLO x 1; soluble LOX 1; SR-EI
Assay Range 0.42 to 10 ng/mL
Assay Sensitivity 0.045 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 108078
Gene Symbol OLR1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target LOX-1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.