Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse LRIG1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 19 publications
Supplier: R&D Systems AF3688SP
This item is not returnable.
View return policy
Description
LRIG1 Polyclonal specifically detects LRIG1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
LRIG1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
D6Bwg0781e, Img, LIG1, LIG-1 | |
Goat | |
Affinity Purified | |
RUO | |
26018 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
LYOPH | |
Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
P70193 | |
Lrig1 | |
Mouse myeloma cell line NS0-derived recombinant mouse LRIG1 Ala37-Thr794 Accession # P70193 | |
25 μg | |
Primary | |
Detects mouse LRIG1 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) LRIG1 is observed and less than 5% cross-reactivity with rhLRIG3 is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction