Learn More
Mouse LRIG1 PE-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | LRIG1 |
| Applications | Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | R-PE |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Gene Accession No. | P70193 |
| Gene Alias | D6Bwg0781e, Img, LIG1, LIG-1 |
| Gene Symbols | Lrig1 |
| Host Species | Goat |
| Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse LRIG1 Ala37-Thr794 Accession # P70193 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.