Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse LRP-6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2960
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2960 100 μg
MAB2960SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2960 Supplier R&D Systems Supplier No. MAB2960
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

LRP-6 Monoclonal specifically detects LRP-6 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen LRP-6
Applications Western Blot
Classification Monoclonal
Clone 290913
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O88572
Gene Alias ADCAD2, FLJ90062, FLJ90421, low density lipoprotein receptor-related protein 6, low-density lipoprotein receptor-related protein 6, LRP6, LRP-6
Gene Symbols LRP6
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse LRP-6 Met1-Pro1368 Accession # O88572
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4040
Test Specificity Detects mouse LRP-6 in direct ELISAs and Western blots. In direct ELISAs, this antibody shows approximately 50 - 60% cross-reactivity with recombinant human (rh) LRP-6 and no cross-reactivity with rhLDL R.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.