Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse LRRC32/GARP Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB62291G10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB62291G10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB62291G10 Supplier R&D Systems Supplier No. FAB62291G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

LRRC32/GARP Monoclonal specifically detects LRRC32/GARP in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen LRRC32/GARP
Applications Flow Cytometry
Classification Monoclonal
Clone 725226
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. NP_001106850
Gene Alias D11S833E, D11S833Eleucine-rich repeat-containing protein 32, GARP, GARPGarpin, Garpin, Glycoprotein A repetitions predominantgarpin, leucine rich repeat containing 32
Gene Symbols LRRC32
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse LRRC32/GARP, Ile18-Asn628, Accession # NP_001106850
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 2615
Test Specificity Detects mouse LRRC32/GARP in direct ELISAs. In direct ELISAs, approximately 50% cross-reactivity with recombinant human (rh) LRRC32 is observed and no cross-reactivity with rhLRRC3, rhLRRC4, or rhNGL-3/LRRC4B is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.