Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse LRRC32/GARP Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. FA62291R100 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA62291R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA62291R100 Supplier R&D Systems Supplier No. FAB62291R100UG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rat Monoclonal Antibody

LRRC32/GARP Monoclonal specifically detects LRRC32/GARP in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen LRRC32/GARP
Applications Flow Cytometry
Classification Monoclonal
Clone 725226
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. NP_001106850
Gene Alias D11S833E, D11S833Eleucine-rich repeat-containing protein 32, GARP, GARPGarpin, Garpin, Glycoprotein A repetitions predominantgarpin, leucine rich repeat containing 32
Gene Symbols LRRC32
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse LRRC32/GARP Ile18-Asn628 Accession # NP_001106850
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2615
Test Specificity Detects mouse LRRC32/GARP in direct ELISAs. In direct ELISAs, approximately 50% cross-reactivity with recombinant human (rh) LRRC32 is observed and no cross-reactivity with rhLRRC3, rhLRRC4, or rhNGL-3/LRRC4B is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Type Antibody
Isotype IgG1
Product Line R&D Systems
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.