Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse LRRC32/GARP Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB62291S100UG
This item is not returnable.
View return policy
Description
LRRC32/GARP Monoclonal specifically detects LRRC32/GARP in Mouse samples. It is validated for Flow Cytometry.Specifications
LRRC32/GARP | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_001106850 | |
LRRC32 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse LRRC32/GARP Ile18-Asn628 Accession # NP_001106850 | |
RUO | |
2615 | |
Mouse | |
IgG1 |
Flow Cytometry | |
725226 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
D11S833E, D11S833Eleucine-rich repeat-containing protein 32, GARP, GARPGarpin, Garpin, Glycoprotein A repetitions predominantgarpin, leucine rich repeat containing 32 | |
Rat | |
100 μg | |
Primary | |
Detects mouse LRRC32/GARP in direct ELISAs. In direct ELISAs, approximately 50% cross-reactivity with recombinant human (rh) LRRC32 is observed and no cross-reactivity with rhLRRC3, rhLRRC4, or rhNGL-3/LRRC4B is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction