Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Ly-6G (Gr-1) Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB10371G100UG
This item is not returnable.
View return policy
Description
Ly-6G/Ly-6C Monoclonal specifically detects Ly-6G/Ly-6C in Mouse samples. It is validated for Flow Cytometry.Specifications
Ly-6G (Gr-1) | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
GR, Gr-1, Ly-6, Ly6G | |
Ly-6G transfected EL-4J cell line, Accession # P35461 | |
100 μg | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
1A8 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P35461 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Detects mouse Ly-6G in Flow Cytometry. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction