Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Ly-6G (Gr-1) Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 9 publications
$200.50 - $479.50
Specifications
Antigen | Ly-6G (Gr-1) |
---|---|
Clone | 1A8 |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Flow Cytometry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10371SP
![]() |
R&D Systems
MAB10371SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10371100
![]() |
R&D Systems
MAB10371100 |
100 μg |
Each for $479.50
|
|
|||||
Description
Ly-6G/Ly-6C Monoclonal specifically detects Ly-6G/Ly-6C in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
Ly-6G (Gr-1) | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Supernatant | |
Mouse | |
P35461 | |
Ly-6G transfected EL-4J cell line Accession # P35461 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Ly-6G in Flow Cytometry. |
1A8 | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Rat | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
GR, Gr-1, Ly-6, Ly6G | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title