Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Ly-6G/Ly-6C (Gr-1) Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 36 publications
$227.50 - $2070.00
Specifications
Antigen | Gr-1/Ly-6G |
---|---|
Clone | RB6-8C5 |
Dilution | Flow Cytometry 0.25 μg/10^6 cells, Immunohistochemistry 3-25 μg/mL, Immunoprecipitation, Immunocytochemistry 8-25 μg/mL, CyTOF-ready |
Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1037SP
![]() |
R&D Systems
MAB1037SP |
25 μg |
Each for $227.50
|
|
|||||
MAB1037100
![]() |
R&D Systems
MAB1037100 |
100 μg |
Each for $561.50
|
|
|||||
MAB1037500
![]() |
R&D Systems
MAB1037500 |
500 μg |
Each for $2,070.00
|
|
|||||
Description
Gr-1/Ly-6G Monoclonal specifically detects Gr-1/Ly-6G in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, CyTOF-ready.Specifications
Gr-1/Ly-6G | |
Flow Cytometry 0.25 μg/10^6 cells, Immunohistochemistry 3-25 μg/mL, Immunoprecipitation, Immunocytochemistry 8-25 μg/mL, CyTOF-ready | |
Monoclonal | |
Rat | |
Mouse | |
C6orf23, G6D, Ly-6C, LY6-D, Ly6G, Ly-6G, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25 | |
Ly6g | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
RB6-8C5 | |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, CyTOF | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
58530 | |
The immunogen for this antibody is Ly-6G (Gr-1, Gr1). | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects Ly-6G/Ly-6C (Gr-1). Weak cross-reactivity with Ly-6C is observed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title