Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Ly-6G/Ly-6C (Gr-1) Biotinylated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems BAM1037
This item is not returnable.
View return policy
Description
Gr-1/Ly-6G Monoclonal specifically detects Gr-1/Ly-6G in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation.Specifications
Gr-1/Ly-6G | |
Monoclonal | |
Biotin | |
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
Ly6g | |
The immunogen for this antibody is Ly-6G (Gr-1, Gr1). | |
250 μg | |
Primary | |
Detects mouse Ly-6G/Ly-6C (Gr-1). Weak cross-reactivity with Ly-6C is observed. | |
Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
RB6-8C5 | |
Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation | |
C6orf23, G6D, Ly-6C, LY6-D, Ly6G, Ly-6G, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
58530 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction