Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Lymphotoxin-alpha/TNF-beta Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB749
This item is not returnable.
View return policy
Description
Lymphotoxin-alpha/TNF-beta Monoclonal specifically detects Lymphotoxin-alpha/TNF-beta in Mouse samples. It is validated for Western Blot.Specifications
Lymphotoxin-alpha/TNF-beta | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
LT, LTA, LT-alpha, Lymphotoxin alpha, lymphotoxin alpha (TNF superfamily, member 1), Lymphotoxinalpha, tnfb, TNF-beta, TNFBlymphotoxin-alpha, TNFSF1, TNFSF1B, TNFSF1TNF-beta, tumor necrosis factor beta, Tumor necrosis factor ligand superfamily member 1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4049 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Western Blot | |
168717 | |
Western Blot 1 ug/mL | |
P09225 | |
LTA | |
E. coli-derived recombinant mouse Lymphotoxin-alpha/TNF-beta Accession # P09225 | |
500 μg | |
Primary | |
Detects mouse Lymphotoxin-alpha/TNF-beta in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) TNF-beta , rhTNF-alpha , rmTNF-alpha , rrTNF-alpha , or rcrTNF-alpha is observed. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction