Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Lymphotoxin beta R/TNFRSF3 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF1008
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF1008 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF1008 Supplier R&D Systems Supplier No. BAF1008
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Lymphotoxin beta R/TNFRSF3 Polyclonal specifically detects Lymphotoxin beta R/TNFRSF3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen Lymphotoxin beta R/TNFRSF3
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL
Gene Accession No. P50284
Gene Alias CD18, D12S370, ltbetar, LT-BETA-R, LTBR, lymphotoxin B receptor, lymphotoxin beta receptor (TNFR superfamily, member 3), Lymphotoxin-beta receptor, LymphotoxinbR, TNF RIII, TNF Rrp, TNFCRTNF-RIII, TNFR superfamily, member 3, TNFR2-RP, TNFR3, TNF-R-III, TNFR-RP, TNFRSF3, TNFRSF3TNFR-III, Tumor necrosis factor C receptor, Tumor necrosis factor receptor 2-related protein, tumor necrosis factor receptor superfamily member 3, tumor necrosis factor receptor superfamily, member 3, Tumor necrosis factor receptor type III
Gene Symbols LTBR
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Lymphotoxin beta R/TNFRSF3 Ser28-Pro218 Accession # P50284
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4055
Test Specificity Detects Lymphotoxin beta R/TNFRSF3 in Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human (rh) LT beta R is observed and less than 1% cross-reactivity with recombinant mouse (rm) 4-1BB, rmCD27, rmCD30, rmCD40, rmEDAR, rmFas, rmGITR, rmOPG, rmRANK, rmTNF RI, and rmTNF RII is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.