Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse LYVE-1 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 27 publications
$212.05 - $2171.79
Specifications
| Antigen | LYVE-1 |
|---|---|
| Clone | 223322 |
| Concentration | LYOPH |
| Dilution | Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction, Immunocytochemistry 8-25 ug/mL, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2125SP
![]() |
R&D Systems
MAB2125SP |
25 μg |
Each for $212.05
|
|
|||||
MAB2125100
![]() |
R&D Systems
MAB2125100 |
100 μg |
Each for $591.67
|
|
|||||
MAB2125500
![]() |
R&D Systems
MAB2125500 |
500 μg |
Each for $2,171.79
|
|
|||||
Description
LYVE-1 Monoclonal specifically detects LYVE-1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF-ready.Specifications
| LYVE-1 | |
| LYOPH | |
| Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q8BHC0 | |
| 10894 | |
| BaF/3 mouse pro-B cell line transfected with mouse LYVE-1 Ala24-Thr234 Accession # Q8BHC0 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse LYVE-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse CD44 or recombinant human LYVE-1 is observed. |
| 223322 | |
| Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE1, LYVE-1XLKD1, XLKD1 | |
| LYVE1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title