Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse M-Cadherin/Cadherin-15 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7677SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7677SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7677SP Supplier R&D Systems Supplier No. MAB7677SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

M-Cadherin/Cadherin-15 Monoclonal specifically detects M-Cadherin/Cadherin-15 in Mouse samples. It is validated for Immunocytochemistry.

Specifications

Antigen M-Cadherin/Cadherin-15
Applications Immunocytochemistry
Classification Monoclonal
Clone 800516
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P33146
Gene Alias cadherin 15, M-cadherin (myotubule), cadherin 15, type 1, M-cadherin (myotubule), Cadherin-14, Cadherin-15, cadherin-3, CDH14, CDH14cadherin-14, CDH15, CDH3cadherin-15, CDHM, MCAD, M-cadherin, MCadherin, MRD3, Muscle cadherin, muscle-cadherin
Gene Symbols CDH15
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse M-Cadherin/Cadherin-15 Val22-Ala605 Accession # P33146
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1013
Test Specificity Detects mouse M-Cadherin/Cadherin-15 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human M-Cadherin, recombinant mouse (rm) Cadherin-13, or rmP-Cadherin is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.