Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse M-CSF Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 7 publications
Supplier: R&D Systems MAB4161SP
This item is not returnable.
View return policy
Description
M-CSF Monoclonal specifically detects M-CSF in Mouse samples. It is validated for Western Blot, Neutralization.Specifications
M-CSF | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
colony stimulating factor 1 (macrophage), CSF1, CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, MCSF, M-CSF, MCSFlanimostim, MGC31930 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1435 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Western Blot, Neutralization | |
131614 | |
Western Blot 1 ug/mL, Neutralization 3-12 ug/mL | |
P07141 | |
CSF1 | |
E. coli-derived recombinant mouse M-CSF Lys33-Glu262 (predicted) Accession # P07141 | |
25 μg | |
Primary | |
Detects mouse M-CSF in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human M-CSF or recombinant mouse SCF is observed. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction