Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse M-CSF R APC-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB3818A
This item is not returnable.
View return policy
Description
M-CSFR/CD115 Monoclonal specifically detects M-CSFR/CD115 in Mouse samples. It is validated for Flow Cytometry.Specifications
M-CSFR/CD115 | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF1R, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, MCSFR, M-CSFR, Proto-oncogene c-Fms | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1436 | |
Mouse | |
Purified |
Flow Cytometry | |
460615 | |
Flow Cytometry 10 uL/10^6 cells | |
P09581 | |
CSF1R | |
Mouse myeloma cell line NS0-derived recombinant mouse M-CSF R Ala20-Ser511 Accession # P09581 | |
100 Tests | |
Primary | |
Detects mouse M-CSF R in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human M-CSF R, recombinant mouse (rm) PDGF R beta , or rmFlt-3 Ligand is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction