Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse M-CSF R/CD115 Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB3818R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB3818R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB3818R100 Supplier R&D Systems Supplier No. FAB3818R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

M-CSFR/CD115 Monoclonal specifically detects M-CSFR/CD115 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen M-CSFR/CD115
Applications Flow Cytometry
Classification Monoclonal
Clone 460615
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P09581
Gene Alias CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF1R, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, MCSFR, M-CSFR, Proto-oncogene c-Fms
Gene Symbols CSF1R
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse M-CSF R/CD115 Ala20-Ser511 Accession # P09581
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1436
Test Specificity Detects mouse M-CSF R/CD115 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human M-CSF R/CD115, recombinant mouse (rm) PDGF R beta , or rmFlt-3 Ligand is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.