Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse MAdCAM-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF993SP
This item is not returnable.
View return policy
Description
MAdCAM-1 Polyclonal specifically detects MAdCAM-1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MAdCAM-1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
hMAdCAM-1, MACAM1, MAdCAM1, MAdCAM-1, mucosal addressin cell adhesion molecule 1, mucosal addressin cell adhesion molecule-1, mucosal vascular addressin cell adhesion molecule 1 | |
Goat | |
Affinity Purified | |
RUO | |
8174 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.25 ug/mL, Immunohistochemistry 5-15 ug/mL | |
NP_038619 | |
MADCAM1 | |
Mouse myeloma cell line NS0-derived recombinant mouse MAdCAM-1 Gln22-Thr365, Predicted Accession # NP_038619 | |
25 μg | |
Primary | |
Detects mouse MAdCAM-1 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) MAdCAM-1, recombinant mouse (rm) ICAM-1, rmICAM-2, rmICAM-5, rmVCAM-1, and rhICAM-3. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction