Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse MAdCAM-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF993
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF993 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF993 Supplier R&D Systems Supplier No. AF993
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

MAdCAM-1 Polyclonal specifically detects MAdCAM-1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen MAdCAM-1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.25 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. NP_038619
Gene Alias hMAdCAM-1, MACAM1, MAdCAM1, MAdCAM-1, mucosal addressin cell adhesion molecule 1, mucosal addressin cell adhesion molecule-1, mucosal vascular addressin cell adhesion molecule 1
Gene Symbols MADCAM1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse MAdCAM-1 Gln22-Thr365, Predicted Accession # NP_038619
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8174
Test Specificity Detects mouse MAdCAM-1 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) MAdCAM-1, recombinant mouse (rm) ICAM-1, rmICAM-2, rmICAM-5, rmVCAM-1, and rhICAM-3.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.