Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse MAG/Siglec-4a Fc Chimera Protein
Click to view available options
Quantity:
100 μg
Description
The Recombinant Mouse MAG/Siglec-4a Fc Chimera Protein is derived from NS0. The Recombinant Mouse MAG/Siglec-4a Fc Chimera Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 4099 |
| Molecular Weight (g/mol) | Observed M.W.: 104-124kDa, reducing conditions, Theoretical M.W.: 82kDa (monomer) |
| Quantity | 100 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse MAG/Siglec-4a protein, (N-terminus) Mouse MAG/Siglec-4a (Gly20-Pro516) Accession &Num; P20917 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Gene Alias | GMAsialic acid binding Ig-like lectin 4A, myelin associated glycoprotein, myelin-associated glycoprotein, sialic acid-binding immunoglobulin-like lectin 4A, Siglec4a, SIGLEC-4A, S-MAG |
| Common Name | MAG/Siglec-4a |
| Gene Symbol | Mag |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction