Learn More
Mouse MARCO Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 3 publications
Supplier:  R&D Systems AF2956SP
Catalog No. AF2956SP
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
MARCO Polyclonal specifically detects MARCO in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
MARCO
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
macrophage receptor with collagenous structureSCARA2Scavenger receptor class A member 2, member 2, SCARA2
Goat
Affinity Purified
RUO
8685
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot
LYOPH
Western Blot 0.25 ug/mL
Q60754
MARCO
Mouse myeloma cell line NS0-derived recombinant mouse MARCO Gln70-Ser518 Accession # Q60754
25 μg
Primary
Detects mouse MARCO in direct ELISAs and Western blots.
Mouse
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only