Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse MARCO Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $491.00
Specifications
| Antigen | MARCO |
|---|---|
| Clone | 2359A |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB29561SP
![]() |
R&D Systems
MAB29561SP |
25 μg |
Each for $200.50
|
|
|||||
MAB29561100
![]() |
R&D Systems
MAB29561100 |
100 μg |
Each for $491.00
|
|
|||||
Description
MARCO Monoclonal specifically detects MARCO in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| MARCO | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| macrophage receptor with collagenous structureSCARA2Scavenger receptor class A member 2, member 2, SCARA2 | |
| Mouse myeloma cell line, NS0-derived mouse MARCO protein, Gln70-Ser518, Accession # Q60754 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2359A | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| Q60754 | |
| 8685 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title