Learn More
Mouse MARCO Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | MARCO |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Clone | 579511 |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 0.25 ug/106 cells, CyTOF-ready |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. |
| Gene Accession No. | Q60754 |
| Gene Alias | macrophage receptor with collagenous structureSCARA2Scavenger receptor class A member 2, member 2, SCARA2 |
| Host Species | Rat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.