Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse MARCO Fluorescein-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB2956F
This item is not returnable.
View return policy
Description
MARCO Monoclonal specifically detects MARCO in Mouse samples. It is validated for Flow Cytometry.Specifications
MARCO | |
Monoclonal | |
Fluorescein | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
macrophage receptor with collagenous structureSCARA2Scavenger receptor class A member 2, member 2, SCARA2 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
8685 | |
Mouse | |
Purified |
Flow Cytometry | |
579511 | |
Flow Cytometry 10 uL/10^6 cells | |
Q60754 | |
MARCO | |
Mouse myeloma cell line NS0-derived recombinant mouse MARCO Gln70-Ser518 Accession # Q60754 | |
100 Tests | |
Primary | |
Detects mouse MARCO in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction