Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Mast Cell Protease-1/Mcpt1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5146SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5146SP 25 μg
MAB5146 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB5146SP Supplier R&D Systems Supplier No. MAB5146SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Mast Cell Protease-1/Mcpt1 Monoclonal specifically detects Mast Cell Protease-1/Mcpt1 in Mouse samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen Mast Cell Protease-1/Mcpt1
Applications Western Blot
Classification Monoclonal
Clone 285008
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P11034
Gene Alias AV080368, Mast Cell Protease1, Mcp1, Mcp-1, MMPC-1
Gene Symbols Mcpt1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Mast Cell Protease-1/Mcpt1 Ile21-Lys246 Accession # P11034
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17224
Test Specificity Detects mouse Mast Cell Protease-1/Mcpt1 in direct ELISAs and Western blots. Detects rat Mast Cell Protease-1/Mcpt1 in Simple Western. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) MCP-6, -7, or -11 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.