Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Mast Cell Protease-1/Mcpt1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5146
This item is not returnable.
View return policy
Description
Mast Cell Protease-1/Mcpt1 Polyclonal specifically detects Mast Cell Protease-1/Mcpt1 in Mouse samples. It is validated for Western Blot, Simple Western.Specifications
Mast Cell Protease-1/Mcpt1 | |
Polyclonal | |
Unconjugated | |
P11034 | |
Mcpt1 | |
Mouse myeloma cell line NS0-derived recombinant mouse Mast Cell Protease-1/Mcpt1 Glu19-Leu246 Accession # P11034 | |
100 μg | |
Primary | |
Detects mouse Mast Cell Protease-1/Mcpt1 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant mouse (rm) Mcpt6, rmMcpt7, and rmMcpt11 is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
AV080368, Mast Cell Protease1, Mcp1, Mcp-1, MMPC-1 | |
Sheep | |
Affinity Purified | |
RUO | |
17224 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction