Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Mast Cell Protease-11/Prss34 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB2857
This item is not returnable.
View return policy
Description
Mast Cell Protease-11/Prss34 Monoclonal specifically detects Mast Cell Protease-11/Prss34 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Mast Cell Protease-11/Prss34 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Q80UR4 | |
Prss34 | |
Mouse myeloma cell line NS0-derived recombinant mouse Mast Cell Protease-11/Prss34 Met20-Ser318 Accession # Q80UR4 | |
100 μg | |
Primary | |
Detects mouse Mast Cell Protease-11/Prss34 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) MCP-1, rmMCP-6, or rmMCP-7 is observed. | |
Mouse | |
Purified |
Western Blot, Immunoprecipitation | |
451421 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EOS, Mast Cell Protease11, MCP-11, MCPT11, Prss34 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
260429 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction