Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Mast Cell Protease-6/Mcpt6 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3736
This item is not returnable.
View return policy
Description
Mast Cell Protease-6/Mcpt6 Polyclonal specifically detects Mast Cell Protease-6/Mcpt6 in Mouse samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.Specifications
Mast Cell Protease-6/Mcpt6 | |
Polyclonal | |
Unconjugated | |
P21845 | |
Goat | |
Affinity Purified | |
RUO | |
17229 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunoprecipitation, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 3 ug/mL | |
Tpsb2 | |
Mouse myeloma cell line NS0-derived recombinant mouse Mast Cell Protease-6/Mcpt6 Ala22-Ser276 Accession # P21845 | |
100 μg | |
Primary | |
Detects Mast Cell Protease-6/Mcpt6 in direct ELISAs and Western blots. In direct ELISAs, approximately 35% cross-reactivity with recombinant human (rh) TPS-1 and rhTPS-B2 is observed and less then 10% cross-reactivity with recombinant mouse MCP-7 is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction