Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Matrilin-3 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF3357
This item is not returnable.
View return policy
Description
MATN3 Polyclonal antibody specifically detects MATN3 in Mouse samples. It is validated for Western Blot.Specifications
MATN3 | |
Polyclonal | |
Biotin | |
AAH71224 | |
NDUFC2 | |
Mouse myeloma cell line NS0-derived recombinant mouse Matrilin-3 Ala35-Arg481 Accession # AAH71224 | |
50 μg | |
Primary | |
Detects mouse Matrilin-3 in Western blots. In Western blots, approximately 50% cross-reactivity with recombinant human Matrilin-3 is observed and less than 5% cross-reactivity with recombinant mouse (rm) Matrilin-2 and rmMatrilin-4 is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
DIPOA, EDM5, HOA, matrilin 3, matrilin-3, Matrilin3, NDUFC2, OADIP, OS2 | |
Goat | |
Affinity Purified | |
RUO | |
4148 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction