Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Melanocortin-3 R/MC3R Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB3737V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB3737V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB3737V100 Supplier R&D Systems Supplier No. FAB3737V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Melanocortin-3 R/MC3R Monoclonal specifically detects Melanocortin-3 R/MC3R in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Melanocortin-3 R/MC3R
Applications Flow Cytometry
Classification Monoclonal
Clone 345404
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Alias BMIQ9, MC3, MC3R, MC3-R, melanocortin 3 receptor, melanocortin receptor 3, Melanocortin3R, Melanocortin-3R, OB20, Obesity quantitative trait locus, OQTL
Gene Symbols MC3R
Host Species Rat
Immunogen Y3 rat myeloid cell line transfeced with mouse Melanocortin 3R/MC3R
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4159
Test Specificity Detects Melanocortin 3R/MC3R in direct ELISAs and Western blots. Stains mouse Melanocortin 3R/MC3R-transfected cells but not irrelevant transfectants.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.