Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Melanocortin-3 R/MC3R Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3737
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3737 100 μg
MAB3737SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB3737 Supplier R&D Systems Supplier No. MAB3737
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Melanocortin-3 R/MC3R Monoclonal specifically detects Melanocortin-3 R/MC3R in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen Melanocortin-3 R/MC3R
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 345404
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias BMIQ9, MC3, MC3R, MC3-R, melanocortin 3 receptor, melanocortin receptor 3, Melanocortin3R, Melanocortin-3R, OB20, Obesity quantitative trait locus, OQTL
Gene Symbols MC3R
Host Species Rat
Immunogen Y3 rat myeloid cell line transfeced with mouse Melanocortin 3R/MC3R
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4159
Test Specificity Detects Melanocortin 3R/MC3R in direct ELISAs and Western blots. Stains mouse Melanocortin 3R/MC3R-transfected cells but not irrelevant transfectants.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.