Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse MEPE/OF45 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6150
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6150 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6150 Supplier R&D Systems Supplier No. AF6150
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

MEPE/OF45 Polyclonal specifically detects MEPE/OF45 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen MEPE/OF45
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/mL
Gene Accession No. AAK70342
Gene Alias extracellular phosphoglycoprotein with ASARM motif (bone), matrix extracellular phosphoglycoprotein, OF45, Osteoregulin
Gene Symbols MEPE
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse MEPE/OF45 Ala17-Asp433 Accession # AAK70342
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56955
Test Specificity Detects mouse MEPE/OF45 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human MEPE/OF45 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.