Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Meprin alpha Subunit/MEP1A Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | Meprin alpha Subunit/MEP1A |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Description
Meprin alpha Subunit/MEP1A Polyclonal specifically detects Meprin alpha Subunit/MEP1A in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Meprin alpha Subunit/MEP1A | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
bA268F1.1 (meprin A alpha (PABA peptide hydrolase)), EC 3.4.24, EC 3.4.24.18, endopeptidase-2, MEP1A, meprin A subunit alpha, meprin A, alpha (PABA peptide hydrolase), N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha, PABA peptide hydrolase, PPH alpha, PPHA | |
MEP1A | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
NP_032611 | |
4224 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Meprin alpha Subunit/MEP1A Val34-Arg615 Accession # NP_032611 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Meprin alpha Subunit/MEP1A in direct ELISAs and Western blots. In direct ELISAs, less than 20% cross-reactivity with recombinant human MEP1A is observed and less than 5% cross-reactivity with recombinant mouse MEP1B is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title