Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Mer Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody has been used in 3 publications

$227.50 - $2150.00

Specifications

Antigen Mer
Clone 108921
Dilution Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB591SP
GREENER_CHOICE
View Documents
R&D Systems
MAB591SP
25 μg
Each for $227.50
Only null left
Add to Cart
 
MAB591100
GREENER_CHOICE
View Documents
R&D Systems
MAB591100
100 μg
Each for $585.00
Only null left
Add to Cart
 
MAB591500
GREENER_CHOICE
View Documents
R&D Systems
MAB591500
500 μg
Each for $2,150.00
Only null left
Add to Cart
 
Description

Description

Mer Monoclonal specifically detects Mer in Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications

Specifications

Mer
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL
Monoclonal
Rat
Mouse
Q60805
10461
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Mer Glu23-Phe498 Accession # Q60805
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects mouse Mer in direct ELISAs and Western blots. Does not cross-react with recombinant human Mer, recombinant mouse (rm) Axl, or rmDtk.
108921
Western Blot, Immunocytochemistry
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
c-Eyk, C-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MerTK, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer
MERTK
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.